Learn More
DWK Life Sciences DURAN™ Sedimentation Cone, Imhoff type, non-graduated
For adjustment and printing.
Brand: DWK Life Sciences 214005402
Code : NEW
Additional Details : Weight : 6.07000kg
Description
DURAN™ sedimentation vessels according to Imhoff are frequently used for the analysis of water quality.
- According to Imhoff without graduation
- Without adjustment and printing
Specifications
1000 mL | |
For the analysis of water quality | |
Borosilicate Glass 3.3 |