missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
Silicone Septum Seal for piercing
£98.40 - £808.00
Specifications
For Use With (Application) | Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments |
---|---|
Material | Silicone |
Quantity | 100/Pk. |
Autoclavable | Autoclavable |
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | ||||||
13177374
|
DWK Life Sciences
292461002 |
GL 45 |
£808.00
Pack of 100 |
In Stock
Log in to see stock. |
||||||
13187164
|
DWK Life Sciences
292460803 |
GL 32 |
£139.00
Pack of 100 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||||
11707198
|
DWK Life Sciences
292460503 |
GL14 |
£98.40
Pack of 100 |
In Stock
Log in to see stock. |
||||||
11717198
|
DWK Life Sciences
292460906 |
GL 25 |
£127.00
Pack of 100 |
In Stock
Log in to see stock. |
||||||
11787198
|
DWK Life Sciences
292460606 |
GL 18 |
£169.00
Pack of 100 |
In Stock
Log in to see stock. |
||||||
Description
Suitable for PBT open topped screw cap with central aperture Heat resistance: 130°C (steam) and 200°C (dry heat). Silicon is peroxide catalysed.
- Compatible with open topped GL screw caps with central aperture
- Compatible with GL bottles
- Peroxide catalysed silicone
Specifications
Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments | |
Silicone | |
100/Pk. | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap