missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Single way Stopcock, complete with hollow key with glass handle

To be used for higher tightness requirements.

Brand:  DWK Life Sciences 286458607

 View more versions of this product

Product Code. 15824798

  • £1340.00 / Pack of 50
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

DURAN™ single way stopcocks complete with hollow DURAN™ cock plug are well suited for work with temperature changes and higher tightness requirements.

  • Ground shut-off valve sleeve with hollow glass plug with glass handle
  • High tightness
  • Taper 1:10
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5 mm
Borosilicate Glass (3.3)
12.5 NS
50/Pk.
8 mm
Autoclavable
1.5 mm
When working with temperature changes and higher tightness requirements; Installation in glass apparatus and devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers