Learn More
DWK Life Sciences DURAN™ Standard Socket, double, finished
For a rigid, self-supporting connection.
£216.00 - £265.00
Specifications
Quantity 50/Pk.
Thickness (Metric) Wall 1.8 mm
View More Specs
Products 2
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15971611
View Documents
DWK Life Sciences
286569808
18 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15844838
View Documents
DWK Life Sciences
286571608
22 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form C. It primarily serves the purpose of saving material.
  • According to DIN 12249 Form C
  • Glass article with two sleeves enables material savings
  • Very low leakage rate
  • Minimized grease requirement
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
50/Pk.
1.8 mm