missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Stirred Bottle Reactor, PP / PTFE / PEEK / Stainless Steel Contact surfaces

Easy-to-use GL 45 lab bottle based reactor system with tubing connectors for use with magnetic stirrers.

£307.00 - £319.00

Specifications

For Use With (Application) Simple, low cost, bioreactor; Simple continuous flow stirred reactor; Mixing liquids, samples or reagents; Photo-bioreactor
Material Polypropylene, PTFE, PEEK, Stainless Steel
Quantity 1 Ea.
Thread Size (Metric) GL 45
Autoclavable Autoclavable
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10642360
View Documents
DWK Life Sciences
1200395
500 mL
£307.00
Each
In Stock
to see stock.
Add to basket
 
10631429
View Documents
DWK Life Sciences
1200396
1000 mL
£319.00
Each
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
 
Description

Description

Self contained bottle stirring system ideal for simple mixing processes. Stirrer shaft can be adjusted to the optimal position in either a 500 or 1000 mL DURAN™ GL 45 bottles. Use with standard magnetic stirrers up to 500 rpm. Significantly improved mixing performance compared to standard magnetic stir bars. Two GL 14 port connector cap is compatible with the modular DURAN™ connection systems; tubing with 1.6 mm to 6.0 mm bores can be used to add or remove liquid or gas. Fully autoclavable. Parts in contact with media conform to FDA requirements. Supplied with or without bottle.

  • The variable stirrer shaft can be adjusted to the optimal position in either 500 or 1000 ml DURAN™ GL 45 bottles
  • Compatible with 1.6 to 6.0 mm outer diameter semi-rigid tubing for addition or removal of liquids or gas
  • Use with standard magnetic stirrers up to 500 rpm and allows a clearly improved mixing
  • Fully compatible with the DURAN™ connection systems
Specifications

Specifications

Simple, low cost, bioreactor; Simple continuous flow stirred reactor; Mixing liquids, samples or reagents; Photo-bioreactor
1 Ea.
Autoclavable
Polypropylene, PTFE, PEEK, Stainless Steel
GL 45
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Stirred Bottle Reactor, PP / PTFE / PEEK / Stainless Steel Contact surfaces

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.