Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
For use with ground DURAN™ single way stopcocks.
£344.00 - £530.00
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Price | Quantity & Availability | |||||
15844518
|
DWK Life Sciences
286036209 |
24.0 NS |
£439.00
Pack of 25 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15834518
|
DWK Life Sciences
286036106 |
21.5 NS |
£344.00
Pack of 25 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15824528
|
DWK Life Sciences
286038407 |
10.0 NS |
£407.00
Pack of 50 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
15854518
|
DWK Life Sciences
286036303 |
29.2 NS |
£530.00
Pack of 25 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
Description
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
- With groove, polished
- The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
- Solid glass tap plugs
- Taper 1:10
Specifications
Borosilicate Glass (3.3) | |
Yes |