Learn More
DWK Life Sciences DURAN™ Stopcock Key, threaded, long for double bore, ground
Product Code. p-7189491 Shop All DWK Life Sciences Products
Click to view available options
Closure Size:
14.5 NS
18.8 NS
Unit Size:
Pack of 50
Product Code. 15864518
Brand: DWK Life Sciences 286036706
Only null left
Add to basket
Add to basket
2-way tap plug with thread.
Ground DURAN™ patent tap plugs have 2 almost parallel inclined bores.
  • With thread, ground
  • 2-way tap plug with 2 almost parallel inclined holes
  • massive tap plug
TRUSTED_SUSTAINABILITY
Specifications
Plug Bore 2.5 mm
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
Closure Size 14.5 NS
No. per Pack 50/Pk.
For Use With (Application) Installation in glass apparatus and devices