Learn More
DWK Life Sciences DURAN™ Test Tube, with beaded rim or straight rim
Test tubes are universal cylindrical laboratory glassware with a round bottom that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
£46.45 - £136.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With | Heating of small amounts; Pretreatments; Storage and transport of small amounts of liquid |
Rimmed | Straight Rim |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10508121
|
DWK Life Sciences
261310809 |
6 mL |
£48.50
Pack of 100 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10612781
|
DWK Life Sciences
261311308 |
16 mL |
£69.85
Pack of 100 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10716811
|
DWK Life Sciences
261312107 |
21 mL |
£79.25
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
10393411
|
DWK Life Sciences
261312304 |
32 mL |
£94.10
Pack of 100 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10403161
|
DWK Life Sciences
261312801 |
40 mL |
£136.00
Pack of 100 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
10577731
|
DWK Life Sciences
261310106 |
2 mL |
£49.00
Pack of 100 |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
10445411
|
DWK Life Sciences
261310303 |
4 mL |
£46.45
Pack of 100 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10211721
|
DWK Life Sciences
261311608 |
17 mL |
£78.05
Pack of 100 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10038170
|
DWK Life Sciences
261313309 |
55 mL |
£72.00
Pack of 50 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
10100551
|
DWK Life Sciences
261313609 |
70 mL |
£79.95
Pack of 50 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10068180
|
DWK Life Sciences
261313806 |
100 mL |
£126.00
Pack of 50 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10538311
|
DWK Life Sciences
261311102 |
8 mL |
£54.40
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10493151
|
DWK Life Sciences
261312604 |
34 mL |
£102.00
Pack of 100 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11670029
|
DWK Life Sciences
261311205 |
9 mL |
£63.55
Pack of 100 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Description
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Thick-walled and therefore mechanically very resistant
- Available with beaded rim or straight rim
- With round bottom
- Fired-on, highly durable white ceramic
Specifications
Borosilicate Glass (3.3) | |
Heating of small amounts; Pretreatments; Storage and transport of small amounts of liquid | |
Straight Rim | |
Autoclavable |