missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks

Especially made for reactions and distillation and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks are manufactured from durable DURAN™ glass.

Brand:  Lenz Laborglasinstrumente™ 03314137

Product Code. 10588081

  • £36.35 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Distillation flasks acc. to DIN 12394 with two angled necks at 20°
  • Reaction flasks acc. to DIN 12392 with two parallel necks at 90°
  • With inscription labels
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

DIN 12394
Borosilicate Glass, Duran Glass
1 Each
Round
100 mL
Three Neck
NS 29/32
64 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.