missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Duran Tubing Reaction Vessels with Thermostatic Jacket, Withdrawal Valve
Glass reaction vessel with thermostatic jacket and withdrawal valve
Brand: Lenz Laborglasinstrumente™ 06030258
This item is not returnable.
View return policy
Description
DURAN™ tubing, cylindrical shape with graduation, laboratory flange LF and withdrawal valve. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.Specifications
500 mL | |
295 mm | |
Reaction vessel | |
Connectors: GL18 |
Borosilicate Glass, Duran Glass | |
Thermostatic jacket, withdrawal valve | |
Cylindrical |