Learn More
DWK Life Sciences DURAN™ Twin-Neck Round Bottom Flask, with standard ground joint, angled side neck
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
£43.75 - £73.95
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volumen sizes 100 mL (NS 24/29 and NS 29/32), 250 mL (NS 24/29), 500 mL (NS 24/29) and 1000 mL (NS 24/29) are not according to DIN ISO |
No. per Pack | 1/Pk. |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
10777481
|
DWK Life Sciences
241832707 |
100 mL | NS 29/32 |
£50.80
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10700493
|
DWK Life Sciences
241833703 |
250 mL | NS 29/32 |
£43.75
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10717681
|
DWK Life Sciences
241834605 |
500 mL | NS 29/32 |
£57.15
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10395431
|
DWK Life Sciences
241835601 |
1000 mL | NS 29/32 |
£56.45
Each |
Estimated Shipment: 03-12-2024
Log in to see stock. |
||||
10598981
|
DWK Life Sciences
241836306 |
2000 mL | NS 29/32 |
£73.95
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
||||
10597841
|
DWK Life Sciences
241832604 |
100 mL | NS 24/29 |
£59.70
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10394951
|
DWK Life Sciences
241834408 |
500 mL | NS 24/29 |
£66.20
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10345531
|
DWK Life Sciences
241833609 |
250 mL | NS 24/29 |
£61.55
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
10408772
|
DWK Life Sciences
241835404 |
1000 mL | NS 24/29 |
£57.70
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||||
Description
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
- Tight closure due to standard ground joint
- angled side necks
- Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
- The geometry permits very uniform heating
- Fired-on, highly durable white print
Specifications
Borosilicate Glass (3.3) | |
Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volumen sizes 100 mL (NS 24/29 and NS 29/32), 250 mL (NS 24/29), 500 mL (NS 24/29) and 1000 mL (NS 24/29) are not according to DIN ISO | |
1/Pk. | |
Autoclavable |