Learn More
DWK Life Sciences DURAN™ Watch Glass Dish, fused rim
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
Brand:  DWK Life Sciences 213216607
54.55 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10202121
  • £63.60 / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
  • Reduced risk of injury due to fused rim
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass
250 mm
Borosilicate Glass (3.3)
1/Pk.