Learn More
DWK Life Sciences DURAN™ Woulff Bottle, 3 standard ground necks
The Woulff bottle is used in vacuum apparatus and serves to collect, clean and/or dry gases.
Brand:  DWK Life Sciences 247097306
 View more versions of this product
Product Code. 10510072
  • ONLINE EXCLUSIVE PRICE
    £292.24
    £269.79 / Each
    Save £22.45
    7% Off
Only null left
Estimated Shipment: 10-12-2025
Add to basket
Description
Description
Woulff''s bottles are part of vacuum apparatus, in which they are used on the one hand as collecting vessels for gas generation or for cleaning and/or drying gases. As part of a vacuum apparatus, Woulff''s bottles are suitable for vacuum applications and often also used as an intermediate vessel to prevent spontaneous drop-off of the vacuum and the associated liquid suction back into the vacuum pump.
  • Tight closure due to standard ground joint
  • Vacuum resistant due to the wall thickness and geometry
  • Three standard ground joints for the import or export of gases
  • Optimum safety for working under pressure or vacuum
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5000 mL
Borosilicate Glass (3.3)
185 mm
Collecting and dissolving gases in liquids; Production of gases; Safe working under vacuum; In series: Purification of a gas
1/Pk.
Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Woulff Bottle, 3 standard ground necks > Neck NS 34/35, 5000 mL