Learn More
DWK Life Sciences DURAN™ YOUTILITY™ Laboratory Bottle, GL 45
The smart ergonomic YOUTILITY™ laboratory bottle system helps to make laboratory work easier and safer.
£57.95 - £117.00
Specifications
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sampling; Culture vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
---|---|
Material | Borosilicate Glass (3.3) |
Quantity | 4/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15243375
|
DWK Life Sciences
218814452 |
500 mL |
£81.70
Pack of 4 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
15253375
|
DWK Life Sciences
218815457 |
1000 mL |
£117.00
Pack of 4 |
In Stock
Log in to see stock. |
|||||
15233375
|
DWK Life Sciences
218812854 |
125 mL |
£57.95
Pack of 4 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
15223375
|
DWK Life Sciences
218813653 |
250 mL |
£65.75
Pack of 4 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Description
The specially shaped gripping zones on both sides of the bottle permit easier and safer handling. The slimmer YOUTILITY™ bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators. The bottles can be conveniently stacked horizontally on their sides. A pre-defined labelling area is compatible with the dedicated self-adhesive YOUTILITY™ bottle labels. Sequentially sized bottles 125, 250, 500 and 1000 ml. The glass bottles are moulded from the tried and tested DURAN™ borosilicate 3.3, a pharmacopoeial Type 1 neutral glass. DURAN™ glass offers a very good chemical resistance and high temperature resistance.
- Easier and safer handling with the specially shaped gripping zones on both sides of the bottle
- Significantly faster opening or closing with the shortened bottle thread which remains fully compatible with GL 45 closures and other accessories
- The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands
- The pre-defined labelling areas are compatible with the dedicated DURAN™ self-adhesive YOUTILITY™ bottle labels for unambiguous bottle identification
- The slimmer bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
- The tried and tested cap plug sealing with sealing lip system ensures a liquid tight seal
- Horizontal stacking is possible
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Autoclaving of media and reagents; Preparation of media and reagents; Sampling; Culture vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir | |
4/Pk. | |
Autoclavable |
Borosilicate Glass (3.3) | |
GL 45 |