missing translation for 'onlineSavingsMsg'
Learn More

Dwyer Inclined Manometer, 3 in. WC

This Inclined Manometer offers pressure measurement for stationary and portable applications.

Brand:  Dwyer 68062-58

Product Code. 11716118

  • £79.15 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ±3% accuracy.
  • Gauge fluid included.
  • Inclined liquid manometers are capable of pressure measurements above and below atmospheric as well as differential pressure measurements.
  • Economically priced, compact and available in both stationary and portable configurations.
  • Construction is simple with virtually indestructible molded white styrene-acrylonitrile housing, indicating tube and fluid wells, molded ABS knobs and zero adjust plunger, shock mounted glass level vial and leak proof O ring seals.
  • These manometers are very easy to use - simply fill the reservoir, adjust fluid level to zero, connect the tubing, and the gauge is ready for operation.
  • Curved inclined-vertical tube of model 68062-58 provides higher ranges with more easily read increments at low readings and is excellent for general purpose use.
Specifications

Specifications

CE Compliance
Gauge Fluid
0 to 3 in. H2O
4.7 mm Barb
10 psi
Pressure measurement for stationary and portable applications
Styrene-acrylonitrile
0.19 in. Barb
±3% Full-scale
Manometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.