Learn More
Fisherbrand™ Dycem™ Polymeric Compound Non-slip Mats
Provides a secure surface that anchors items such as cups, plates, tools, and a variety of other objects.
Brand:  Fisherbrand™ 2047/9
 View more versions of this product
Product Code. 12856445
  • £16.30 / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Can be washed with soap and water
  • Latex-free
Specifications
Specifications
Blue
320 x 260 mm
Dychem Polymeric Compound