missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Dye Concentrate
Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Dark Red
Yellow/Green
pH Range:
5.1 to 5.3
8
8.6 to 8.8
Unit Size:
Each
Beschreibung
- Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters.
- Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult
- Won’t harm the environment—nontoxic and biodegradable
- Easy-to-use color in rapidly diffusing dye identifies dispersion patterns, flow rates, seepage and more Noncarcinogenic fluorescent red dye is ideal for yellowed or heavily algaed water
- Brighten murky ponds with blue dye
- Minimum detectable concentration is 1 ppm
Spezifikation
Spezifikation
| Color | Yellow/Green |
| pH Range | 8 |
| For Use With (Application) | Squirt-top dispensing |
| Quantity | 1/Pk. |
| Type | Dye Tracer |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur