missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Epitope Tag Antibody, Agarose, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Bio-Techne NB600-351
Additional Details : Weight : 0.00970kg
Description
DYKDDDDK Epitope Tag Polyclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunoprecipitation.Specifications
DYKDDDDK Epitope Tag | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunoprecipitation | |
DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag | |
To produce DYKDDDDK Epitope Tag Antibody, Goats were immunized with ECS, Enterokinase Cleavage Site (xxxDDDDK), conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4C. Do not freeze. | |
Epitope Tag |
Western Blot, Immunoprecipitation | |
Agarose | |
PBS with 0.09% Sodium Azide | |
Goat | |
1.01 kDa | |
0.1 mg | |
ECS Epitope Tags (DDDDK), Epitope Tags | |
DYKDDDDK Epitope Tag Antibody reacts with FLAG. | |
IgG |
For Research Use Only