Learn More
DYKDDDDK Tag Monoclonal Antibody (FG4R), DyLight™ 800 4X PEG, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191878D800
Code : NEW
Additional Details : Weight : 0.00900kg
Description
MA1-91878-D800 detects FLAG tag from various samples. MA1-91878-D800 has been successfully used for fluorescent detection of FLAG tagged proteins in Western Blotting. Similar epitope as FLAG tag from Sigma. DyLight 800 4X PEG has an excitation/emission of 783/797 nm.
DYKDDDDK Tag Monoclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Western Blot.Specifications
DYKDDDDK Tag | |
Monoclonal | |
1 mg/mL | |
MES with proprietary stabilizer and no preservative | |
Mouse | |
Protein A | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Western Blot | |
FG4R | |
DyLight 800 4X PEG | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
Synthetic peptide (DYKDDDDK) coupled to KLH. | |
50 μL | |
Primary | |
Antibody | |
IgG1 |