Learn More
Thermo Scientific™ DyLight™ 550 NHS Ester
Tienda Thermo Scientific ProductosDescripción
Thermo Scientific™ Amine-Reactive DyLight Dyes are NHS ester-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as molecular probes for cellular imaging and other fluorescence detection methods.
Highlights:
Brightness: high-fluorescence intensity provides outstanding sensitivity and requires less conjugate
Photostability: exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4pi microscopy)
Instrument compatibility: emission spectra match the principal output wavelengths of common fluorescence instrumentation
Especificaciones
Especificaciones
| Quantity | 1 mg |
| Product Type | Dye |
| Solubility | DMF (Dimethylformamide) |
| Content And Storage | Store desiccated at -20°C. |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Label or Dye | DyLight™ 550 |
| Label Type | DyLight Fluors |
| Product Line | DyLight |
| Emission | 576 |
| Mostrar más |
Seguridad y manipulación
- DyLight 550 NHS Ester,1 mg.
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : DyLight Reactive Dyes
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.