missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyLight™ Antibody Labeling Kits

Covalently label antibodies or proteins with DyLight dyes for use in immunofluorescence, flow cytometry, western blot analysis, and high content analysis.

Brand:  Thermo Scientific™ 84535

285.12 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 11834574

  • £432.00 / 1 set
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Invitrogen Dylight Antibody Labeling kits provide everything needed to label 1 mg of antibody or protein with amine-reactive Dylight dye and subsequent free dye removal. Choose from a variety of Dylight fluorophores across the spectrum to covalently label your antibody or protein. Labeled antibodies are typically ready to use in 90 minutes with minimal hands-on time. The kit contains all necessary components to perform three separate labeling reactions using 1 mg of IgG or similar quantities of other proteins.

Invitrogen DyLight Antibody Labeling kits enable the creation of fluorescent antibodies that are covalently linked to different DyLight fluorophores. The fluorescently labeled antibodies can be used in a wide variety of laboratory applications, including immunofluorescence microscopy, IHC, western blotting, ELISA, flow cytometry, or in vitro and in vivo fluorescent detection strategies.

DyLight Antibody Labeling kits contain an NHS ester-activated derivative of the high-performance DyLight dye that can be used to fluorescently label antibodies and other proteins for cellular imaging and other fluorescence detection methods. The DyLight dye is an amine-reactive dye and is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds.

DyLight Antibody Labeling kits contain all necessary components to perform three separate labeling reactions and free dye removal using 1 mg of IgG or similar quantities of other proteins.

DyLight dyes exhibit higher fluorescence intensity and photostability than CyDye and LI-COR™ dyes in many applications and remain highly fluorescent over a broad pH range (pH 4–9). Additionally, the water solubility of the DyLight fluorescent dyes allows a high dye-to-protein ratio to be achieved without causing precipitation of conjugates. Finally, the high solubility of these fluorophores permits protein solutions to be added directly to specific amounts of the labeling reagent.

Other advantages of the DyLight Antibody Labeling kits:
· High performance—high fluorescence intensity across a broad pH range
· Multiple colors—10 different dyes across the visible spectrum and extending into the infrared
· Specificity—NHS ester-activated dye labels proteins and other molecules at primary amines (-NH2)
· Optimized procedure—following the standard protocol results in antibodies with excellent dye:protein ratios and recovery rates for optimum activity and fluorescence labeling

We also offer DyLight Microscale Antibody Labeling kits for fast and efficient fluorescent labeling of antibodies for use in fluorescence methods. The DyLight Microscale Antibody Labeling kits contain all necessary components to perform five separate labeling reactions using 100 μg of IgG or similar quantities of other proteins. Both the standard and microscale kit sizes include the amine-reactive DyLight fluorophore in convenient single-use vials as well as purification resin and spin columns for the preparation of ready-to-use conjugate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains 3 vials DyLight NHS Ester, 1 mL Tube Borate Buffer, 5 mL Purification Resin, 6 Spin Columns, and 12 Microcentrifuge Collection Tubes
Store the DyLight NHS Ester at -20°C. Store all other kit components at 4°C.
0.5 to 1 mg
Amine
Far-Red
DyLight™
Labeling Kit
652/672
DyLight™
DyLight 650
Antibodies, Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DyLight Antibody Labeling Kits

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B

Hazard Statement
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : disodium tetraboratedecahydrate, boricacid, DyLight Reactive Dyes

SDS
Documents

Documents

Certificates
Special Offers

Special Offers