Learn More
Applied Biosystems™ Dynabeads™ anti-Salmonella
Shop All Applied Biosystems ProductsDescription
For the selective enrichment of Salmonella directly from pre-enrichment samples using ImmunoMagnetic Separation. Method can be performed manually or can be automated by using a Thermo Scientific KingFisher mL Food Protection Purification System.
Benefits and features
- Increases Sensitivity and positive isolation rate
- Easy IMS Protocol—30 minutes from start to finish
- Shelf stable reagent and low start up costs
This Product Contains: Dynabeads coated with anti-Salmonella antibodies
Sensitivity:
Sensitivity is very high and enables detection of 1 viable organism in 25 g of sample or as few as 100 organisms/mL after pre-enrichment.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2°C to 8°C). |
| Type | Magnetic Beads |
| Prep Scale | Small Volume Microbial Samples |
| Product Line | Dynabeads |
| Quantity | 5 mL |
| Shipping Condition | Room Temperature |
| Target Organism Class | Salmonella spp. |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.