missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Biotin Binder

Product Code. p-200105789
Click to view available options
Quantity:
2 mL
5 mL
Unit Size:
2mL
5mL
This item is not returnable. View return policy

Product Code. 30275945

Brand: Invitrogen™ 11048D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dynabeads Biotin Binder contains 2.8 μm superparamagnetic beads that bind biotinylated antibodies (or other biotinylated ligands), allowing you to isolate cells from any species (e.g., human, mouse, rat, non-human primates, and cell lines).

Dynabeads Biotin Binder contains 2.8 μm superparamagnetic beads that bind biotinylated antibodies (or other biotinylated ligands), allowing you to isolate cells from any species (e.g., human, mouse, rat, non-human primates, and cell lines). Advantages of Dynabeads Biotin Binder:

  • Works with any cell type, from any sample and from any species
  • Fast and efficient depletion or positive isolation of any target cells for molecular downstream assays using a biotinylated primary antibody
  • Obtain pure and untouched cells using a mixture of biotinylated antibodies to deplete unwanted cells

Adaptable for various isolation strategies

The biotinylated primary antibody may be added to the cell sample (indirect technique) or pre-coated onto the Dynabeads Biotin Binder (direct technique). Regardless of which technique is used, when the cell sample and the Dynabeads Biotin Binder are mixed, the beads bind to the target cells. Placing the sample on a magnet separates the bead-bound cells from the rest of the sample. One or more biotinylated antibodies may be used to either deplete the sample of unwanted cells (negative isolation) or to capture specific cells for downstream experiments (positive isolation). For isolation of bead-free cells (e.g., for flow cytometry), we recommend using your biotinylated antibody in combination with the CELLection Biotin Binder Kit.

Useful for many sample types

Using Dynabeads Biotin Binder, you can isolate cells basically from any species, and from many starting samples such as whole blood, cord blood, peripheral blood mononuclear cells (PBMC), buffy coat, mouse spleen/lymph nodes, and tissue digests.

Learn more about Dynabeads products

TRUSTED_SUSTAINABILITY

Specifications

Cell Type All cells from all species
Content And Storage This Product Contains: 2 mL Dynabeads™ coated with recombinant streptavidin.
Color Brown
For Use With (Application) Cell Depletion, Negative Isolation, Positive Isolation
Material Magnetic Polystyrene
Product Type Biotin Binder
No. of Cells Processes ∼8x 10^8 cells total
Output Viability >95%
Product Line Dynabeads
Purity or Quality Grade Research Grade
Quantity 2 mL
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 10^7 PBMCs per isolation
Target Species All species
Diameter (Metric) 2.8 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.