missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Carboxylic Acid for RNA Purification

Dynabeads Carboxylic Acid for RNA Purification are monosized (1 μm), monodispersed, paramagnetic beads characterized by high speed to magnet and high pellet stability.

Brand:  Invitrogen™ 65020D

187.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 17857378

  • £220.00 / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dynabeads Carboxylic Acid for RNA Purification are monosized (1 μm), monodispersed, paramagnetic beads characterized by high speed to magnet and high pellet stability. They are intended for use with Dynabeads RNA Binding Buffer, for simple, scalable, and automation-ready purification of crude RNA produced by in vitro transcription (IVT). The combination is particularly suitable where flexibility and high purity are required, such as during the discovery phase of mRNA vaccine development and following the solid-phase IVT reaction with Dynabeads Streptavidin for In Vitro Transcription.

Features of Dynabeads Carboxylic Acid for RNA Purification include:

  • Efficiency—reuse at least six times to reduce the cost and time in a simple workflow
  • Scalability—swap between manual and automated workflows to match your required scale (micrograms to grams of mRNA) and throughput
  • Quality—ensure safe and flexible supply of magnetic beads to navigate from RNA-based drug discovery to commercial manufacturing without incurring extra time or costs from switching technology or raw material

Crude RNA from an IVT reaction step is mixed with the beads and the buffer is added. The RNA binds to the bead surface and the remaining components of the reaction mixture are removed by applying a magnet and discarding the supernatant.

The bead concentration is 10 mg/mL. They are stored in purified water and may be recycled at least six times. Scripts are available for KingFisher automated purification workflows.

The beads are offered here as a stand-alone product. The are also included in the kits below:

  • Dynabeads RNA Purification Kit (Cat. No. 65030D)
  • Dynabeads In Vitro Transcription and RNA Purification Kit (Cat. No. 65034D)

To learn more about other products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics visit: thermofisher.com/dynabeadsmrna

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mg/mL
Dynabeads magnetic beads are supplied in purified water. Store at 2°C to 8°C.
Magnetic Bead
KingFisher™ Sample Purification System, DynaMag™ magnets
Dynabeads™
2 mL
2 mL
Magnetic Polystyrene
Brown
1 μm
Yes
In Vitro Transcription, Synthetic RNA, RNA Purification
Carboxylic Acid Magnetic Bead
Ambient Temperature
ISO9001 and ISO13485
Monosized 1 μm (CV <5%)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.