missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ MyOne™ Silane

Product Code. 12336182
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 12336182

Brand: Invitrogen™ 37002D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An excellent tool for highly predictable and consistent extraction and isolation of nucleic acids from biological samples, following a simple magnetic separation protocol.

An excellent tool for highly predictable and consistent extraction and isolation of nucleic acids from biological samples, following a simple magnetic separation protocol. Please note that this is a stand-alone product containing only Dynabeads™ in a storage buffer. Our products Dynabeads™ SILANE genomic DNA (Cat.no. 370-12D) or Dynabeads™ SILANE viral NA (Cat.no. 370-11D) include the appropriate buffers and protocols you need to extract genomic DNA or viral nucleic acids respectively.

Magnetic particles from alternative suppliers often have a random size range distribution, surface area and binding capacity. This could compromise the reproducibility of your results. With Dynabeads™ MyOne™ SILANE you are ensured a higher sensitivity, capacity and performance.

The small 1μm Dynabeads™ MyOne™ SILANE have an optimized silica-like surface chemistry and a high specific surface area, providing efficient kinetics and a high sensitivity in nucleic acid capture. Their uniform size and surface area also ensure reproducible results. The product holds reputable Dynal™ high standards with respect to within- and between-lot reproducibility and automation ability.

Advantages

  • Highly predictable binding per mg of beads
  • High sensitivity, allowing for a low detection limit
  • For isolation of viral DNA/RNA, total RNA, genomic DNA, etc.
  • Isolates both DNA and RNA
  • Quicker than spin columns
  • More cost-effective, yet performs to the same level as spin columns
  • Automation-friendly (slow sedimentation rate + high magnetic mobility)
  • Adjusted to customer requirements on a custom OEM basis

Applications

  • Isolation of total nucleic acids (DNA⁄RNA)from typical biological samples (e.g. serum/plasma, whole blood, etc.)
  • Well suited for automated assays.
  • Dedicated kits have been developed for isolation of viral nucleic acids and genomic DNA (available separately).

Additional Info
Validation and customization (bead, buffer, protocol or format) can be made available on a custom OEM basis. For further information and price quotes for larger volumes of the silica-like Dynabeads™ MyOne™ SILANE, please contact us at: ivd@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 40 mg/mL
Content And Storage Supplied at a concentration of 40 mg/mL. 5 mL total volume, sufficient for 96 isolations.

Store at: 2°C to 8°C.
Product Type SILANE
Product Line DYNAL, Dynabeads, MyOne
Quantity 5 mL
Shipping Condition Room Temperature
Surface Functionality Silanol (Silica-Like)
Diameter (Metric) 1 μm

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.