missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Sequencing Clean-Up Kit

Product Code. p-7181028
Click to view available options
Quantity:
1 mL, 1 mL Kit (500 Samples)
5 mL, 5 mL Kit (2500 Samples)
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10592045

Brand: Invitrogen™ 66101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads Sequencing Clean-Up offers a simple one-tube system for purifying DNA sequencing reactions.

The Invitrogen Dynabeads Sequencing Cleanup Kit uses uniform, superparamagnetic beads for efficient magnetic separation and quickly purifies dye-terminator sequencing reactions by removing excess dye terminators, enzymes, and salts. This kit helps deliver exceptional performance for capillary electrophoresis (CE) applications, including all Sanger sequencing–based applications.

Features of the Dynabeads Sequencing Cleanup Kit include:

  • Automation-compatible with 96- and 384-well plate workflows
  • No centrifugation or filtration needed
  • Rapid one-tube protocol completed in just 15 minutes
  • Efficient removal of excess dye terminators resulting in dye blob–free sequencing data
  • Generates sequences with a high number of Phred 20 bases
  • Beads made with exceptional quality, backed by over 30 years of f knowledge and experience
  • Consistent, reliable, and efficient Sanger sequencing reaction cleanup due to the uniform shape, size, and high binding capacity of Dynabeads magnetic beads
  • Custom solutions available, tailored to meet specific research needs
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C, This product contains Dynabeads™ in binding buffer
Store at 2°C to 8°C.
For Use With (Application) DNA and RNA Purification and Analysis, DNA Extraction, Sequencing Reaction Purification, Sequencing
Includes Dynabeads in binding buffer
Isolation Technology Magnetic Bead
Sample Type Sequencing Reactions, Enzymatic Reactions, Sequencing Reactions
Quantity 1 mL, 1 mL Kit (500 Samples)
High-throughput Compatibility High-throughput Compatible
Product Line DYNAL, Dynabeads
Product Type Sequencing Clean-Up Kit
Shipping Condition Room Temperature
Target Sequencing Reactions, DNA from Enzymatic Reactions (e.g. PCR)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.