Learn More
Invitrogen™ DynaMag™-PCR Magnet
The MAGnify™ ChIP system utilizes a novel magnet that is compatible with 0.2mL PCR strip tubes.
Brand: Invitrogen™ 492025
275.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.05000kg
Description
The MAGnify™ ChIP system utilizes a novel magnet that is compatible with 0.2mL PCR strip tubes. This magnet is critical for enabling the use of reduced starting cell numbers and volumes for immunoprecipitation and washing steps. This novel magnetic separator allows you to achieve results quickly and easily while offering the freedom to simultaneously perform multiple ChIP assays in PCR tubes with a multi-channel pipettor.Specifications
Store magnet at room temperature. | |
DynaMag™ | |
DynaMag™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.