Learn More
Abeba™ Dynamic 37310 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37310/46
 View more versions of this product
Product Code. 15630848
  • £60.05 / 1 pair
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
37310
White
46
Polyurethane