Learn More
Abeba™ Dynamic 37313 Shoes
Product Code. 15610878
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15610878
Brand: Abeba™ 37313/46
Only null left
Add to basket
Add to basket
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather white/red
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White
Material Leather
Size 46
Sole Features Slip-resistant
Sole Material Polyurethane
Style 37313