Learn More
Abeba™ Dynamic 37313 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
£60.05
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, E, FO SRC |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15600868
|
Abeba™
37313/35 |
35 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15610868
|
Abeba™
37313/36 |
36 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15620868
|
Abeba™
37313/37 |
37 |
£60.05
1 pair |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
15630868
|
Abeba™
37313/38 |
38 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15640868
|
Abeba™
37313/39 |
39 |
£60.05
1 pair |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
15650868
|
Abeba™
37313/40 |
40 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15660868
|
Abeba™
37313/41 |
41 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15670868
|
Abeba™
37313/42 |
42 |
£60.05
1 pair |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
15680868
|
Abeba™
37313/43 |
43 |
£60.05
1 pair |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
15690868
|
Abeba™
37313/44 |
44 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15600878
|
Abeba™
37313/45 |
45 |
£60.05
1 pair |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
15610878
|
Abeba™
37313/46 |
46 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15620878
|
Abeba™
37313/47 |
47 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15630878
|
Abeba™
37313/48 |
48 |
£60.05
1 pair |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/red
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Polyurethane |
White | |
Slip-resistant | |
37313 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Abeba™ Dynamic 37313 Shoes