Learn More
Abeba™ Dynamic 37343 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 37343/46
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15630988
  • £60.94 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber with breathable textile inlays
  • Black/Yellow
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Microfiber
Slip-resistant
37343
Black, Yellow
46
Polyurethane