Learn More
Abeba™ Dynamic 37361 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 37361/36
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Microfiber black
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Microfiber | |
Slip-resistant | |
37361 |
Black | |
36 | |
Polyurethane |