Learn More
Abeba™ Dynamic 7315 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Brand:  Abeba™ 7315/45
Additional Details : Weight : 0.98000kg
 View more versions of this product
Product Code. 15632688
  • £53.42 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather
  • Black/brown
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7315
Black
45
Polyurethane