Learn More
ABEBA™ Dynamic Blue Occupational Clogs Ob ESD
Blue dynamic occupational clog with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
Brand: ABEBA™ 37326/39
This item is not returnable.
View return policy
Description
- Breathable, lightweight, comfortable occupational clogs
- Comply to EN ISO 20347: 2012, OB SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Water repellent, blue knitted textile
- Non foldable heel strap, adjustable
- Removable active comfort insole
Specifications
Blue | |
39 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC | |
Clogs | |
OB SRC ESD |