missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DyNAmo HS SYBR Green qPCR Kit
Contains a modified Thermus brockianus DNA polymerase and all the other reagents needed for qPCR.
£487.00 - £2315.00
Specifications
Detection Method | SYBR |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Tbr DNA polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10788298
|
Thermo Scientific™
F410L |
500 Reactions |
£487.00
200 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10206079
|
Thermo Scientific™
F410XL |
2500 Reactions |
£2315.00
2500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Only template DNA and PCR primers need to be added by the user. The modified DNA polymerase in this kit incorporates a non-specific DNA-binding domain that lends physical stability to the polymerase-DNA complex. ROX passive reference dye is also supplied with the kit in a separate tube for instruments that require ROX normalization.
- Strong specificity: DyNAmo HS SYBR Green master mix minimizes the formation of unwanted primer-dimers and extension of non-specifically bound primers during reaction setup
- Highly sensitive detection: Optimized master mix allows reproducible detection of low-copy number templates
- Broad dynamic range: DyNAmo HS SYBR Green qPCR Kit detects as few as one copy of the target gene and achieves a dynamic range of up to eight orders of magnitude
- Convenient to use: 2X master mix, one tube only; ROX passive reference dye provided in a separate vial
Specifications
SYBR | |
High | |
Tbr DNA polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler | |
DyNAmo | |
Gene Expression | |
DNA, dsDNA |
Tube | |
qPCR | |
Standard | |
DyNAmo HS SYBR Green qPCR Kit | |
Real Time PCR SYBR Master Mix | |
2X (DyNAmo HS SYBR Green qPCR Mix), 50X (ROX Dye) |
For Research Use Only. Not for use in diagnostic procedures.