missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) carbonate tetrahydrate, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11337108

Dysprosium(III) carbonate tetrahydrate, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11337108
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11337108

Brand: Thermo Scientific Alfa Aesar 015286.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-35-1
Molecular Formula C3H8Dy2O13
Molecular Weight (g/mol) 577.084
MDL Number MFCD00150229
InChI Key BQRZSEZQNZPPPU-UHFFFAOYSA-H
Synonym dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
PubChem CID 25021600
IUPAC Name dysprosium(3+);tricarbonate;tetrahydrate
SMILES C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]

Specifications

Physical Form Powder
Quantity 50 g
Formula Weight 577.06 (505.03 Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) carbonate tetrahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253632 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number015286.18,015286.09
Lot NumberR26H012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015286.18,015286.09
Lot NumberA03Y025 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015286.18,015286.09
Lot NumberJ14N08 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015286.18,015286.09
Lot NumberU16H031 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number015286.18,015286.09

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Dysprosium(III) carbonate tetrahydrate, 99.9% (REO), Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado