missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11363877

Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11363877
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11363877

Brand: Thermo Scientific Alfa Aesar 035693.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used in high-intensity metal-halide lamps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15474-63-2
Molecular Formula DyI3
Molecular Weight (g/mol) 543.213
MDL Number MFCD00016072
InChI Key RZQFCZYXPRKMTP-UHFFFAOYSA-K
Synonym dysprosium iii iodide, dysprosium triiodide, dysprosium iodide, acmc-20al3a, dysprosium iii ; iodide, dysprosium 3+ triiodide, dysprosium iii iodide, anhydrousShow MoreShow Less
PubChem CID 84931
IUPAC Name dysprosium(3+);triiodide
SMILES [I-].[I-].[I-].[Dy+3]

Specifications

Melting Point 955°C
Color Green to Yellow
Physical Form Flakes
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 543.21
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) iodide, Ultra dry
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP11L014 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number035693.14,035693.03,035693.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.