missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11311069

Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11311069
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11311069

Brand: Thermo Scientific Alfa Aesar 011315.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dysprosium(III) nitrate is used for catalysts. It is used in the preparation of dysprosium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-49-9
Molecular Formula DyH10N3O14
Molecular Weight (g/mol) 438.59
MDL Number MFCD00149683
InChI Key NOTQUFQJAWMLCE-UHFFFAOYSA-N
IUPAC Name dysprosium(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 88.6°C
Color Cream to Yellow
pH 3
Physical Form Crystalline Aggregates
Quantity 100 g
UN Number UN1477
Merck Index 14,3482
Solubility Information Soluble in water.
Formula Weight 438.59 (348.51 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Dysprosium(III) nitrate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.