missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11311079

Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11311079
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11311079

Brand: Thermo Scientific Alfa Aesar 011319.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Cermets Used As Nuclear Reaction Control Rods

In cermets used as nuclear reaction control rodsDysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts by reacting with acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-87-8
Molecular Formula Dy2O3
Molecular Weight (g/mol) 373.00
MDL Number MFCD00010986
InChI Key GEZAXHSNIQTPMM-UHFFFAOYSA-N
IUPAC Name didysprosium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Dy+3].[Dy+3]

Specifications

Melting Point 2340°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 373
Sensitivity Hygroscopic
Appearance White powder or lump
Density 7.81^2^7
Chemical Name or Material Dysprosium(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ23K002 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number011319.30,011319.09,011319.18
Lot NumberT26K040 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number011319.30,011319.09,011319.18
Lot NumberW04F047 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number011319.30,011319.09,011319.18
Lot Number61500167 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011319.30,011319.09,011319.18
Lot Number61300733 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011319.30,011319.09,011319.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.