missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) oxide, REacton™, 99.99% (REO), Thermo Scientific Chemicals

£131.66 - £1724.58

Chemical Identifiers

CAS 1308-87-8
Molecular Formula Dy2O3
Molecular Weight (g/mol) 373.00
MDL Number MFCD00010986
InChI Key GEZAXHSNIQTPMM-UHFFFAOYSA-N
IUPAC Name didysprosium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Dy+3].[Dy+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381069
View Documents
Thermo Scientific Alfa Aesar
011318.14
25 g
£131.66
25g
Please to purchase this item. Need a web account? Register with us today!
11391069
Ver documentos
Thermo Scientific Alfa Aesar
011318.22
100 g
£377.88
100g
Please to purchase this item. Need a web account? Register with us today!
11371069
Ver documentos
Thermo Scientific Alfa Aesar
011318.36
500 g
£1724.58
500g
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Dysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts via reaction with acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones

Identificadores de productos químicos

1308-87-8
373.00
GEZAXHSNIQTPMM-UHFFFAOYSA-N
[O--].[O--].[O--].[Dy+3].[Dy+3]
Dy2O3
MFCD00010986
didysprosium(3+) trioxidandiide

Especificaciones

2340°C
Powder
MFCD00010986
Insoluble in water.
[O--].[O--].[O--].[Dy+3].[Dy+3]
373.00
99.99%
Hygroscopic
(REO)
1308-87-8
Dy2O3
14,3482
GEZAXHSNIQTPMM-UHFFFAOYSA-N
didysprosium(3+) trioxidandiide
373
Reacton™
7.81 g/mL at 27°C
Dysprosium(III) oxide
Videos
Seguridad y manipulación

Seguridad y manipulación

missing translation for 'einecsNumber' : 215-164-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Product Certifications

RUO – Research Use Only