missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11331609

Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11331609
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11331609

Brand: Thermo Scientific Alfa Aesar 012257.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dysprosium pieces, 99.9%, CAS Number-7429-91-6, atom, powder, pieces, foil, 3n, disprosio, chips, 3n, ion, ingot, ingot, 3n, ingot reo, 100 g, (REO), CHEBI:33377, Silver, Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S., 231-073-9, Danger

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Silver
UN Number UN3178
Quantity 25 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Dysprosium pieces, Distilled dendritic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP17D015 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number012257.14,012257.22,012257.06
Lot NumberF4258 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012258.14,012257.14,012257.22,012257.06
Lot NumberK11W044 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012258.14,012257.14,012257.22,012257.06
Lot NumberA11L02 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012258.14,012257.14,012257.22,012257.06
Lot NumberP17D015 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012258.14,012257.14,012257.22,012257.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.