missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium rod, 6.35mm (0.25in) dia, 99.7% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog Number 11333808
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
This item is not returnable. View return policy
7429-91-6
Dy
162.50
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
23912
CHEBI:33377
dysprosium
[Dy]
This item is not returnable. View return policy
7429-91-6
Dy
162.50
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
23912
CHEBI:33377
dysprosium
[Dy]

Form: Approx. 0.27g/mm, Cast rod, diameter is approximate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray to Silver
Physical Form Cast Rod
Quantity 10 mm
Assay Percent Range 99.7% (Metals basis excluding Ta)
Odor Odorless
Appearance Gray to silver rod
Chemical Name or Material Dysprosium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'