missing translation for 'onlineSavingsMsg'
Learn More

DZIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen DZIP1
Concentration 0.13 mg/mL
Applications Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16852164
View Documents
Proteintech
13779-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16842164
View Documents
Proteintech
13779-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May participate in spermatogenesis via its interaction with DAZ.
Specifications

Specifications

DZIP1
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Mouse, Human
Q86YF9, Q8BMD2
22873, 66573
DZIP1 Fusion Protein Ag4772
Primary
-20°C
DZIP1
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
DAZ interacting protein 1, DAZ interacting protein 1/2, DZIP, DZIP1, DZIP2, DZIPt1, KIAA0996, RP11 23E3.3, Zinc finger protein DZIP1
Dzip1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates