Learn More
3M™ E-A-R™ Classic™ Earplugs
Product Code. p-8006569
Click to view available options
Corded:
Corded
Uncorded
Noise Reduction Rating (NRR):
28db
Unit Size:
200 pairs
250 pairs
Product Code. 12354749
Brand: 3M™ 7000052844
Only null left
Add to basket
Add to basket
Provides excellent hearing protection and all day comfort. 3M E-A-R Classic Earplug offer either 28 or 29dB.
Made from a soft, energy-absorbing polymer foam that provides excellent hearing protection and all day comfort. Ideal for use where noise is a problem, either in work or leisure activities.
  • Firm material with slow recovery time helps achieve good fit
  • Special cylindrical shape that fits most ear canals and provides reliable seal
  • Low equilibrium pressure helps reduce pressure in ear canal thus increasing comfort and wearability
  • One size fits majority of wearers
  • Excellent sound attenuation characteristics
  • Sweat and humidity resistant which helps improve comfort
  • Ideally suited for use in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Certifications/Compliance CE Approved
Quantity 200/Pk.
Noise Reduction Rating (NRR) 28db
Corded Corded
Product Type Classic Earplug
Style Roll-down