Learn More
3M™ E-A-R™ Classic™ Probed Test Plugs Product Code.: 15719787
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
3M™ E-A-R™ Classic™ Probed Test Plugs
Product Code. 15719787
Color: Yellow
Click to view available options
Color: Yellow
Product Code. 15719787
Brand: 3M™ 7000052882
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
Add to basket
3M™ E-A-R™ Classic™ Probed Test Plugs equipped with a probe tube for fit testing using the 3M™ E-A-Rfit™ Validation System.
  • These test earplugs are modified with a probe tube for use during earplug fit testing with the 3M™ E-A-Rfit™ Validation System
  • Test plugs can not be used for hearing protection
  • Probed test earplug for use with 3M™ E-A-Rfit™ Validation System
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Size Small
Packaging Type Polybag
Pairs per Package 50 Pairs/Box
Corded Uncorded
Plug Shape Cylinder
Dielectric No
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
3M™ E-A-R™ Classic™ Probed Test Plugs >