Learn More
3M™ E-A-R™ Classic™ Small Earplugs
Click to view available options
:
Size: Small
Description
- The E-A-R™ Classic™ Small Earplugs may be used for protection against moderate to high noise environments, providing an SNR of 29 dB as fully tested and approved against EN 352-2:2020.
- Uncorded
Specifications
Specifications
| Size | Small |
| Noise Reduction Rating (NRR) | 29 dB |
| Corded | Uncorded |
| Plug Shape | Cylinder |
| Disposable | Single-use |
| Metal Detectable | No |
| Dielectric | No |
| Mounting | No |