Learn More
3M™ E-A-R™ Classic™ SuperFit™ Earplug
Click to view available options
Color: Yellow, Orange
Description
- Earplug with a orange fitting ring so its easy to when the earplug is correctly fitted
- When the orange color is no longer visible, the best fit is achieved
- Smaller in diameter and length to fit smaller ear canals or those looking for slightly lower attenuation
- The Earplugs are moisture resistant and compatible with other PPE
Specifications
Color | Yellow, Orange |
Pairs per Package | 200 Pairs/Box |
Noise Reduction Rating (NRR) | 33 dB |
Corded | Uncorded |
Product Type | Earplugs |
Dielectric | No |