Learn More
3M™ E-A-R™ E-A-Rsoft™ Yellow Neons Probed Test Plugs
Click to view available options
Color: Yellow
Description
- These test earplugs are modified with a probe tube for use during earplug fit testing with the 3M™ E-A-Rfit™ Validation System
- Test plugs can not be used for hearing protection
- Probed test earplug for use with 3M™ E-A-Rfit™ Validation System
Specifications
Specifications
Color | Yellow |
Size | Universal |
Packaging Type | Polybag |
Pairs per Package | 50 Pairs/Box |
Noise Reduction Rating (NRR) | 33 dB |
Plug Shape | Tapered |
Product Type | Ear Plugs |
Dielectric | No |
Material | Polyurethane |
Product Content Correction
Product Title
3M™ E-A-R™ E-A-Rsoft™ Yellow Neons Probed Test Plugs >
Spot an opportunity for improvement?Share a Content Correction