Learn More
3M™ E-A-R™ Tri-Flange™ Earplugs
Click to view available options
Cord Material:
Cloth
Vinyl
Description
- Earplugs with triple flanges that adapt to the shape of the ear to form multiple seals against noise
- No need to handle the plug tip making them more hygienically acceptable
- Triple-flange design preformed to fit the ear
- Reusable plug is comfortable, hygienic and economical as it can be washed
- Supplied with a cotton or vinyl cord
Specifications
Specifications
Color | Yellow |
Certifications/Compliance | CE Approved |
Cord Material | Cloth |
Pairs per Package | 400 Pairs/Box |
Noise Reduction Rating (NRR) | 29 dB |
Dielectric | No |
Material | Elastomeric Polymer |
Product Content Correction
Product Title
3M™ E-A-R™ Tri-Flange™ Earplugs >
Spot an opportunity for improvement?Share a Content Correction