Learn More
3M™ E-A-R™ Ultrafit™ Ear Plugs, Corded
Product Code. p-8006571
Click to view available options
Color:
Green
Yellow
Yellow, Blue
Noise Reduction Rating (NRR):
14 db
20 db
32 db
Material:
Elastomeric Polymer
Silicone Rubber
TPE
Thermoplastic Elastomer
Quantity:
200/Pk.
50/Pk.
Unit Size:
50 pairs
Pack of 200
Product Code. 12602565
Brand: 3M™ 7000038199
Only null left
Add to basket
Add to basket
3M™ E-A-R™ UltraFit™ Ear Plugs are reusable and have a proven triple-flange design that fits comfortably in most ears. They reduce noise levels by 14-32 dB and can be cleaned easily with soapy water and reused again and again – helping decrease waste.
  • Triple-flange pliable plug effectively fits most ear canal sizes
  • Reduces noise levels by 14-32 dB
  • Ideal for dirty environments as no roll down required, simply push them into your ears for hygienic protection
  • Clean easily with soapy water and reuse again and again, helping decrease waste
  • Removable, reusable cord attachment – convenient if you move in and out of noisy environments
  • Available with a storage case to keep plugs clean and together
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow, Blue
Certifications/Compliance CE Approved
Quantity 50/Pk.
Noise Reduction Rating (NRR) 32 db
Product Type Ear Plugs
Material Elastomeric Polymer